missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Furil, 98%

Catalog No. AAA1538606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538606 5 g
AAA1538614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1538606 Supplier Thermo Scientific Chemicals Supplier No. A1538606

CAS: 492-94-4 | C10H6O4 | 190.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-94-4
Molecular Formula C10H6O4
Molecular Weight (g/mol) 190.154
MDL Number MFCD00003241
InChI Key SXPUVBFQXJHYNS-UHFFFAOYSA-N
Synonym furil, alpha-furil, bipyromucyl, 2,2'-furil, bipryomucyl, di-2-furylglyoxal, difuranylglyoxal, ethanedione, di-2-furanyl, di-2-furanylethanedione, a-furil
PubChem CID 68119
IUPAC Name 1,2-bis(furan-2-yl)ethane-1,2-dione
SMILES C1=COC(=C1)C(=O)C(=O)C2=CC=CO2

Specifications

Melting Point 162°C to 165°C
Quantity 5 g
Beilstein 383882
Formula Weight 190.15
Percent Purity 98%
Chemical Name or Material 2,2'-Furil
EINECSNumber 207-766-7
RTECSNumber LV0580000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only