missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Fluorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. F09335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09335G 5 g
F093325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09335G Supplier TCI America Supplier No. F09335G

Chemical Identifiers

CAS 52721-69-4
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075502
InChI Key RIKUOLJPJNVTEP-UHFFFAOYSA-O
Synonym 2-fluorophenethylamine, 2-2-fluorophenyl ethanamine, 2-2-fluorophenyl ethylamine, benzeneethanamine, 2-fluoro, 2-2-fluoro-phenyl-ethylamine, 2-2-fluorophenyl ethan-1-amine, 2-fluoro-phenethylamine, 2-fluorophenethyl amine, 2-fluorobenzeneethanamine, benzeneethanamine,2-fluoro
PubChem CID 643357
IUPAC Name 2-(2-fluorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CC=C1F

Specifications

Color Colorless
Boiling Point 64°C
Quantity 5 g
UN Number 2735
Formula Weight 139.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2-Fluorophenyl)ethylamine