missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethyl bromide, 98%, stab. with potassium carbonate

Catalog No. AAL0852209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0852209 10 g
AAL0852218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0852209 Supplier Thermo Scientific Chemicals Supplier No. L0852209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 54550-36-6 | C6H13BrO2 | 197.072 g/mol

2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Ethoxyethoxy)ethyl bromide is used as pharmaceutical intermediates.

Solubility
Not miscible with water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 54550-36-6
Molecular Formula C6H13BrO2
Molecular Weight (g/mol) 197.072
MDL Number MFCD00051920
InChI Key UEDIWIFQWRXXJG-UHFFFAOYSA-N
Synonym 1-bromo-2-2-ethoxyethoxy ethane, 2-2-ethoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-ethoxyethane, 1-bromo-2-2-ethoxy-ethoxy-ethane, acmc-1axh5, 1-bromo-3,6-dioxaoctane, 1-2-ethoxyethoxy-2-bromoethane, 1-ethoxy-2-2-bromoethoxy ethane, 1-2-bromoethoxy-2-ethoxy-ethane, 1-bromo 2-2-ethoxyethoxy-ethane
PubChem CID 7015324
IUPAC Name 1-(2-bromoethoxy)-2-ethoxyethane
SMILES CCOCCOCCBr

Specifications

Density 1.299
Boiling Point 86°C to 88°C (12 mmHg)
Refractive Index 1.453
Quantity 10 g
Beilstein 1736603
Solubility Information Not miscible with water.
Formula Weight 197.08
Percent Purity 98%
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl bromide, stabilized with potassium carbonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only