missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethyl acetate, 99%

CAS: 112-15-2 | C8H16O4 | 176.21 g/mol

$231.24 - $231.24

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00041928
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
Synonym carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
PubChem CID 8165
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC257420025
SDS View Documents
Thermo Scientific Chemicals
257420025
2.5 L Glass bottle
Each for $231.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-15-2
176.21
FPZWZCWUIYYYBU-UHFFFAOYSA-N
8165
CCOCCOCCOC(C)=O
C8H16O4
MFCD00041928
carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
2-(2-ethoxyethoxy)ethyl acetate

Specifications

112-15-2
98.5
-25.0°C
1.0100g/mL
98°C
98.5% min. (GC)
C8H16O4
CH3C(O)OCH2CH2OCH2CH2OC2H5
2.5 L
1.01
carbitol acetate, 2-2-ethoxyethoxy ethyl acetate, diethylene glycol monoethyl ether acetate, ektasolve de acetate, karbitolacetat, ethoxydiglycol acetate, glycol ether de acetate, diglycol monoethyl ether acetate, ethanol, 2-2-ethoxyethoxy-, acetate, ethyldiglycol acetate
FPZWZCWUIYYYBU-UHFFFAOYSA-N
2-(2-ethoxyethoxy)ethyl acetate
8165
176.2
Extra Pure
2-(2-Ethoxyethoxy)ethyl acetate
0.2 mg KOH/g max.
100.0
Beige to White
210.0°C to 220.0°C
Authentic
Glass bottle
1.4206 to 1.4226
MFCD00041928
0.2% max.
15, 3134
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, most oils
CCOCCOCCOC(C)=O
176.21
2.8 mPa.s (20°C)
99%
Crystalline Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

EINECSNumber : 203-940-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only