missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethanol, Spectrum™ Chemical

Catalog No. p-7165526
Change view
Click to view available options
Quantity:
500 mL
2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Catalog No. Quantity Packaging
18608146 500 mL Amber Glass Bottle
18608142 2.5 L Glass Bottle
18608143 200 L Drum
3 options

Catalog No. 18608146

Supplier: Spectrum Chemical Mfg Cor E1265500MLGL

Only null left

Chemical Identifiers

CAS 111-90-0

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.