missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethanol, Spectrum™ Chemical

Catalog No. 18608146
Change view
Click to view available options
Quantity:
500 mL
2.5 L
200 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608146 500 mL Glass Bottle
18608142 2.5 L Glass Bottle
18608143 200 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18608146 Supplier Spectrum Chemical Mfg Cor Supplier No. E1265500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 111-90-0

Specifications

CAS Max % 1
Boiling Point 196°C to 197°C (384.8°F to 386.6°F)
Packaging Glass Bottle
Quantity 500 mL
Molecular Weight (g/mol) 134.18
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy)ethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.