missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™ Chemical

Catalog No. 18608148
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608148 4 L Steel Can
18608147 1 L Steel Can
18608144 20 L Steel Can
18608145 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18608148 Supplier Spectrum Chemical Mfg Cor Supplier No. E10224LTGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
IUPAC Name 2-(2-ethoxyethoxy)ethan-1-ol
SMILES CCOCCOCCO

Specifications

Acidity 0.0001
CAS Max % 1
Boiling Point 196°C to 197°C (384.8°F to 386.6°F)
Packaging Steel Can
Quantity 4 L
Percent Purity 99 to 100%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy)ethanol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.