missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor E102220LTMT

 View more versions of this product

Catalog No. 18608144


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

0.0001
1
C6H14O3
XXJWXESWEXIICW-UHFFFAOYSA-N
2-(2-ethoxyethoxy)ethan-1-ol
99 to 100%
111-90-0
Steel Can
20 L
CCOCCOCCO
134.18
Reagent

Chemical Identifiers

111-90-0
134.18
2-(2-ethoxyethoxy)ethan-1-ol
C6H14O3
XXJWXESWEXIICW-UHFFFAOYSA-N
CCOCCOCCO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.