missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dipyrrolylmethane 97.0+%, TCI America™
$281.48 - $980.49
Chemical Identifiers
| CAS | 21211-65-4 |
|---|---|
| Molecular Formula | C9H10N2 |
| Molecular Weight (g/mol) | 146.19 |
| MDL Number | MFCD03931560 |
| InChI Key | PBTPREHATAFBEN-UHFFFAOYSA-N |
| PubChem CID | 140814 |
| ChEBI | CHEBI:36319 |
| IUPAC Name | 2-[(1H-pyrrol-2-yl)methyl]-1H-pyrrole |
| SMILES | C(C1=CC=CN1)C1=CC=CN1 |
Chemical Identifiers
| 21211-65-4 | |
| 146.19 | |
| PBTPREHATAFBEN-UHFFFAOYSA-N | |
| CHEBI:36319 | |
| C(C1=CC=CN1)C1=CC=CN1 |
| C9H10N2 | |
| MFCD03931560 | |
| 140814 | |
| 2-[(1H-pyrrol-2-yl)methyl]-1H-pyrrole |
Specifications
| 21211-65-4 | |
| White-Yellow | |
| C9H10N2 | |
| 1 g | |
| C(C1=CC=CN1)C1=CC=CN1 | |
| 146.19 | |
| CHEBI:36319 | |
| ≥97.0% (GC) | |
| 2,2′-Dipyrrolylmethane |
| 74°C | |
| 120°C | |
| MFCD03931560 | |
| PBTPREHATAFBEN-UHFFFAOYSA-N | |
| 2-[(1H-pyrrol-2-yl)methyl]-1H-pyrrole | |
| 140814 | |
| 146.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator