missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dipyridylamine 99.0+%, TCI America™
$152.08 - $517.87
Chemical Identifiers
| CAS | 1202-34-2 |
|---|---|
| Molecular Formula | C10H9N3 |
| Molecular Weight (g/mol) | 171.203 |
| MDL Number | MFCD00006247 |
| InChI Key | HMMPCBAWTWYFLR-UHFFFAOYSA-N |
| Synonym | 2,2'-dipyridylamine, di 2-pyridyl amine, di pyridin-2-yl amine, 2,2'-bipyridylamine, 2-pyridinamine, n-2-pyridinyl, n-pyridin-2-yl pyridin-2-amine, unii-x9bf664yak, pyridinamine, n-pyridinyl, pyridine, 2,2'-iminodi, 2,2-dipyridylamine |
| PubChem CID | 14547 |
| IUPAC Name | N-pyridin-2-ylpyridin-2-amine |
| SMILES | C1=CC=NC(=C1)NC2=CC=CC=N2 |
Chemical Identifiers
| 1202-34-2 | |
| 171.203 | |
| HMMPCBAWTWYFLR-UHFFFAOYSA-N | |
| 14547 | |
| C1=CC=NC(=C1)NC2=CC=CC=N2 |
| C10H9N3 | |
| MFCD00006247 | |
| 2,2'-dipyridylamine, di 2-pyridyl amine, di pyridin-2-yl amine, 2,2'-bipyridylamine, 2-pyridinamine, n-2-pyridinyl, n-pyridin-2-yl pyridin-2-amine, unii-x9bf664yak, pyridinamine, n-pyridinyl, pyridine, 2,2'-iminodi, 2,2-dipyridylamine | |
| N-pyridin-2-ylpyridin-2-amine |
Specifications
| 1202-34-2 | |
| White-Yellow | |
| C10H9N3 | |
| 5 g | |
| HMMPCBAWTWYFLR-UHFFFAOYSA-N | |
| N-pyridin-2-ylpyridin-2-amine | |
| 14547 | |
| ≥99.0% (GC,T) | |
| 2,2′-Dipyridylamine |
| 94°C | |
| 178°C | |
| MFCD00006247 | |
| 2,2'-dipyridylamine, di 2-pyridyl amine, di pyridin-2-yl amine, 2,2'-bipyridylamine, 2-pyridinamine, n-2-pyridinyl, n-pyridin-2-yl pyridin-2-amine, unii-x9bf664yak, pyridinamine, n-pyridinyl, pyridine, 2,2'-iminodi, 2,2-dipyridylamine | |
| C1=CC=NC(=C1)NC2=CC=CC=N2 | |
| 171.203 | |
| 171.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : UR3545000
TSCA : Yes