missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dipyridylamine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Color | White-Yellow |
| Boiling Point | 178°C |
| Quantity | 5 g |
| Formula Weight | 171.20 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Dipyridylamine |
Safety and Handling
| RTECSNumber | UR3545000 |
| TSCA | Yes |