missing translation for 'onlineSavingsMsg'
Learn More

2,2-Diphenylethanol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 1883-32-5
Molecular Formula C14H14O
Molecular Weight (g/mol) 198.26
MDL Number MFCD00004729
InChI Key NYLOEXLAXYHOHH-UHFFFAOYSA-N
Synonym beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
PubChem CID 74662
IUPAC Name 2,2-diphenylethanol
SMILES C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC188220020
SDS View Documents
Thermo Scientific Chemicals
188220020
2g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC188220100
SDS View Documents
Thermo Scientific Chemicals
188220100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1883-32-5
198.26
NYLOEXLAXYHOHH-UHFFFAOYSA-N
74662
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
C14H14O
MFCD00004729
beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
2,2-diphenylethanol

Specifications

1883-32-5
100.0
>110°C
96% min. (GC)
C14H14O
MFCD00004729
06, II, 640
NYLOEXLAXYHOHH-UHFFFAOYSA-N
2,2-diphenylethanol
74662
97%
96.0
160°C to 162°C (5.0mmHg)
Authentic
Glass bottle
(C6H5)2CHCH2OH
2g
beta-phenylphenethyl alcohol, 2,2-diphenyl ethanol, benzeneethanol, .beta.-phenyl, acmc-20aojg, 2,2-diphenyl-ethanol, 2,2-diphenyl-1-ethanol, 2,2-diphenylethan-1-ol, benzeneethanol, b-phenyl, 2,2-diphenylethanol, ksc539o6l
C1=CC=C(C=C1)C(CO)C2=CC=CC=C2
198.26
198.26
2, 2-Diphenylethanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-543-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only