missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinaphthyl Ether 99.0+%, TCI America™

Catalog No. p-7126446
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D081110G 10 g
D081125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D081110G Supplier TCI America Supplier No. D081110G
Only null left

Chemical Identifiers

CAS 613-80-9
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.33
MDL Number MFCD00015447
InChI Key DZRLNYVDCIYXPG-UHFFFAOYSA-N
PubChem CID 69183
IUPAC Name 2-(naphthalen-2-yloxy)naphthalene
SMILES O(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 105°C
Color White-Yellow
Boiling Point 250°C
Quantity 10 g
Synonym 2,2'-dinaphthyl ether, di-2-naphthyl ether, 2,2'-oxydinaphthalene, naphthalene, 2,2'-oxybis, bis 2-naphthyl ether, 2-naphthyl ether, di-2-naphthylether, 2,2'-dinaphthylether, naphthalene,2,2'-oxybis
Formula Weight 270.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Dinaphthyl Ether