missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2'-Dinaphthyl Ether 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 105°C |
| Color | White-Yellow |
| Boiling Point | 250°C |
| Quantity | 10 g |
| Synonym | 2,2'-dinaphthyl ether, di-2-naphthyl ether, 2,2'-oxydinaphthalene, naphthalene, 2,2'-oxybis, bis 2-naphthyl ether, 2-naphthyl ether, di-2-naphthylether, 2,2'-dinaphthylether, naphthalene,2,2'-oxybis |
| Formula Weight | 270.33 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,2′-Dinaphthyl Ether |
Safety and Handling
| TSCA | No |