missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dinaphthyl ether, 98+%

Catalog No. AAL0256506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0256506 5 g
AAL0256514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0256506 Supplier Thermo Scientific Chemicals Supplier No. L0256506
Only null left

CAS: 613-80-9 | C20H14O | 270.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-80-9
Molecular Formula C20H14O
Molecular Weight (g/mol) 270.33
MDL Number MFCD00015447
InChI Key DZRLNYVDCIYXPG-UHFFFAOYSA-N
PubChem CID 69183
IUPAC Name 2-naphthalen-2-yloxynaphthalene
SMILES O(C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2054097
Synonym 2,2'-dinaphthyl ether, di-2-naphthyl ether, 2,2'-oxydinaphthalene, naphthalene, 2,2'-oxybis, bis 2-naphthyl ether, 2-naphthyl ether, di-2-naphthylether, 2,2'-dinaphthylether, naphthalene,2,2'-oxybis
Formula Weight 270.33
Percent Purity ≥98%
Chemical Name or Material 2,2'-Dinaphthyl ether
EINECSNumber 210-356-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only