missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylvaleric Acid 98.0+%, TCI America™

Catalog No. p-7127228
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23205ML 5 mL
D232025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23205ML Supplier TCI America Supplier No. D23205ML
Only null left

Chemical Identifiers

CAS 1185-39-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00021686
InChI Key ZRYCZAWRXHAAPZ-UHFFFAOYSA-N
Synonym 2,2-Dimethylpentanoic Acid
PubChem CID 14455
IUPAC Name 2,2-dimethylpentanoic acid
SMILES CCCC(C)(C)C(=O)O

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 mL
UN Number 3265
Formula Weight 130.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylvaleric Acid
RTECSNumber YV6487500
TSCA No