missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylsuccinic Anhydride 98.0+%, TCI America™

Catalog No. D44175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44175G 5 g
D441725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44175G Supplier TCI America Supplier No. D44175G

Chemical Identifiers

CAS 17347-61-4
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00022608
InChI Key ACJPFLIEHGFXGP-UHFFFAOYSA-N
Synonym 2,2-dimethylsuccinic anhydride, 3,3-dimethyldihydrofuran-2,5-dione, unii-tl903a7h7v, 2,5-furandione, dihydro-3,3-dimethyl, 2,2-dimethyl succinic anhydride, 2,2-dimethylsuccinicanhydride, dihydro-3,3-dimethylfuran-2,5-dione, 2,2-dimethylsuccinic acid anhydride, dihydro-3,3-dimethyl-2,5-furandione, ksc490q1j
PubChem CID 87067
IUPAC Name 3,3-dimethyloxolane-2,5-dione
SMILES CC1(C)CC(=O)OC1=O

Specifications

Melting Point 30°C
Color White
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Boiling Point 220°C
Formula Weight 128.13
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,2-Dimethylsuccinic Anhydride