missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylpentane 99.0+%, TCI America™

Catalog No. p-7126868
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16731ML 1 mL
D16735ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16731ML Supplier TCI America Supplier No. D16731ML
Only null left

Chemical Identifiers

CAS 590-35-2
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009404
InChI Key CXOWYJMDMMMMJO-UHFFFAOYSA-N
Synonym pentane, 2,2-dimethyl, unii-m82t90qevx, m82t90qevx, acmc-1b0g6, 2,2-dimethylpentane
PubChem CID 11542
IUPAC Name 2,2-dimethylpentane
SMILES CCCC(C)(C)C

Specifications

Color Colorless
Boiling Point 79°C
Quantity 1 mL
UN Number 1206
Formula Weight 100.21
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylpentane
EINECSNumber (2)-0007
TSCA Yes