missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylhexane 98.0+%, TCI America™

Catalog No. D12241ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12241ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12241ML Supplier TCI America Supplier No. D12241ML
Only null left

Chemical Identifiers

CAS 590-73-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009485
InChI Key FLTJDUOFAQWHDF-UHFFFAOYSA-N
PubChem CID 11551
IUPAC Name 2,2-dimethylhexane
SMILES CCCCC(C)(C)C

Specifications

Color Colorless
Boiling Point 106°C
Quantity 1 mL
UN Number 1262
Formula Weight 114.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylhexane
EINECSNumber (2)-0008
TSCA No