missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclopentanone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4541-32-6
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00049164
InChI Key FTGZMZBYOHMEPS-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one
PubChem CID 138286
IUPAC Name 2,2-dimethylcyclopentan-1-one
SMILES CC1(CCCC1=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC399400010
View Documents
Thermo Scientific Chemicals
399400010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399400050
View Documents
Thermo Scientific Chemicals
399400050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4541-32-6
112.17
FTGZMZBYOHMEPS-UHFFFAOYSA-N
138286
CC1(CCCC1=O)C
C7H12O
MFCD00049164
2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one
2,2-dimethylcyclopentan-1-one

Specifications

4541-32-6
100.0
143°C to 145°C
Authentic
Glass bottle
1.4320 to 1.4340 (20°C, 589nm)
MFCD00049164
0.89
FTGZMZBYOHMEPS-UHFFFAOYSA-N
2,2-dimethylcyclopentan-1-one
138286
97%
96.0
0.8900g/mL
32°C
96% min. (GC)
C7H12O
(CH3)2C5H6(=O)
1g
2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one
CC1(CCCC1=O)C
112.17
112.17
2, 2-Dimethylcyclopentanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only