missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclopentanone, 97%

Catalog No. AC399400010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399400010 1 g Glass bottle
AC399400050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399400010 Supplier Thermo Scientific Chemicals Supplier No. 399400010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4541-32-6 | C7H12O | 112.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4541-32-6
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00049164
InChI Key FTGZMZBYOHMEPS-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclopentanone, cyclopentanone, 2,2-dimethyl, cyclopentanone, dimethyl, zlchem 1228, 2,2 dimethylcyclopentanone, 2,2-dimethyl-cyclopentanone, 2,2-dimethylcyclo pentanone, 2,2-dimethylcyclo-pentanone, 2,2-dimethyl-cyclopentan-1-one
PubChem CID 138286
IUPAC Name 2,2-dimethylcyclopentan-1-one
SMILES CC1(CCCC1=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Density 0.8900g/mL
Boiling Point 143°C to 145°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C5H6(=O)
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.89
Formula Weight 112.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2, 2-Dimethylcyclopentanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only