missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylbutyric Acid 96.0+%, TCI America™

Catalog No. D2429500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2429500ML 500 mL
D242925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2429500ML Supplier TCI America Supplier No. D2429500ML

Chemical Identifiers

CAS 595-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00004200
InChI Key VUAXHMVRKOTJKP-UHFFFAOYSA-N
Synonym 2,2-dimethylbutyric acid, butanoic acid, 2,2-dimethyl, 2,2-dimethyl-butanoic acid, 2,2-dimethyl butyric acid, 2,2-dimethyl butanoic acid, unii-ay606cn05o, butyric acid, 2,2-dimethyl, dimethyl ethyl acetic acid, 2,2-dimethyl-butyric acid, .alpha.,.alpha.-dimethylbutyric acid
PubChem CID 11684
ChEBI CHEBI:38649
IUPAC Name 2,2-dimethylbutanoic acid
SMILES CCC(C)(C)C(=O)O

Specifications

Color Yellow
Boiling Point 186°C
Quantity 500 mL
UN Number 3265
Formula Weight 116.16
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethylbutyric Acid
EINECSNumber (2)-0608
TSCA No