missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dimethylbiphenyl 95.0+%, TCI America™

Catalog No. D17171G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17171G 1 g
D17175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17171G Supplier TCI America Supplier No. D17171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 605-39-0
Molecular Formula C14H14
Molecular Weight (g/mol) 182.27
MDL Number MFCD00048075
InChI Key ABMKWMASVFVTMD-UHFFFAOYSA-N
Synonym 2,2'-dimethylbiphenyl, 1,1'-biphenyl, 2,2'-dimethyl, o,o'-bitolyl, 2,2'-ditolyl, 2,2'-bitolyl, 2,2'-dimethyl-1,1'-biphenyl, 1-methyl-2-2'-methylphenyl benzene, 1-methyl-2-2-methylphenyl benzene, bitoluene, bitolyl
PubChem CID 11797
IUPAC Name 2,2'-dimethyl-1,1'-biphenyl
SMILES CC1=CC=CC=C1C1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 259°C
Quantity 1 g
Formula Weight 182.27
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2′-Dimethylbiphenyl