missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(Dimethylamino)ethoxy]ethanol 98.0+%, TCI America™
Supplier: TCI America D1756500ML
Specifications
| 2-[2-(Dimethylamino)ethoxy]ethanol | |
| Yellow | |
| C6H15NO2 | |
| 500 mL | |
| YSAANLSYLSUVHB-UHFFFAOYSA-N | |
| 2-[2-(dimethylamino)ethoxy]ethan-1-ol | |
| 74348 | |
| ≥98.0% (GC) |
| 1704-62-7 | |
| 95°C | |
| MFCD00059602 | |
| Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
| CN(C)CCOCCO | |
| 133.19 | |
| 133.19 | |
| Liquid |
Chemical Identifiers
| 1704-62-7 | |
| 133.19 | |
| YSAANLSYLSUVHB-UHFFFAOYSA-N | |
| 74348 | |
| CN(C)CCOCCO |
| C6H15NO2 | |
| MFCD00059602 | |
| Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
| 2-[2-(dimethylamino)ethoxy]ethan-1-ol |
Safety and Handling
RTECSNumber : KK6825000
TSCA : Yes