missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Dimethylamino)ethoxy]benzoic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC51301DA

 View more versions of this product

Catalog No. CC51301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-[2-(Dimethylamino)ethoxy]benzoic acid
94.5
95%
C11H15NO3
1g
JZLYEZGHBQFRLU-UHFFFAOYSA-N
2-[2-(dimethylamino)ethoxy]benzoic acid
6484659
95%
206261-66-7
100.0
Amber Glass Bottle
MFCD07801180
2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
CN(C)CCOC1=CC=CC=C1C(=O)O
209.245
209.24

Chemical Identifiers

206261-66-7
209.245
JZLYEZGHBQFRLU-UHFFFAOYSA-N
6484659
CN(C)CCOC1=CC=CC=C1C(=O)O
C11H15NO3
MFCD07801180
2-2-dimethylamino ethoxy benzoic acid, 2-2-dimethylamino-ethoxy-benzoic acid, 2-2-dimethylaminoethoxy benzoic acid, 2-2-dimethylaminoethyloxy benzoic acid, 2-carboxy-beta-dimethylamino phenetole, 2-2-n,n-dimethylamino ethoxy benzoic acid, 2-2-n,n-dimethylamino ethyl oxy-benzoic acid
2-[2-(dimethylamino)ethoxy]benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions