missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione 98.0+%, TCI America™

Catalog No. D20955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20955G 5 g
D209525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20955G Supplier TCI America Supplier No. D20955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15231-78-4
Molecular Formula C12H12O4
Molecular Weight (g/mol) 220.224
MDL Number MFCD00006636
InChI Key NTUAHNQWHPQAMB-UHFFFAOYSA-N
Synonym 1,3-dioxane-4,6-dione, 2,2-dimethyl-5-phenyl, 5-phenylmeldrum's acid, 5-phenyl-meldrum's acid, acmc-1c8jq, 2.2-dimethyl-5-phenyl-1.3-dioxane-4.6-dione, 1,3-dioxane-4,6-dione,2,2-dimethyl-5-phenyl, 2,2-dimethyl-5-phenyl-1,3 dioxane-4,6-dione
PubChem CID 84848
IUPAC Name 2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione
SMILES CC1(OC(=O)C(C(=O)O1)C2=CC=CC=C2)C

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 220.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethyl-5-phenyl-1,3-dioxane-4,6-dione