missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-4-pentenoic Acid 98.0+%, TCI America™
$127.21 - $505.03
Chemical Identifiers
| CAS | 16386-93-9 |
|---|---|
| Molecular Formula | C7H12O2 |
| Molecular Weight (g/mol) | 128.17 |
| MDL Number | MFCD00012304 |
| InChI Key | BGUAPYRHJPWVEM-UHFFFAOYSA-N |
| Synonym | 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g |
| PubChem CID | 140065 |
| IUPAC Name | 2,2-dimethylpent-4-enoic acid |
| SMILES | CC(C)(CC=C)C(O)=O |
Chemical Identifiers
| 16386-93-9 | |
| 128.17 | |
| BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
| 140065 | |
| CC(C)(CC=C)C(O)=O |
| C7H12O2 | |
| MFCD00012304 | |
| 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
| 2,2-dimethylpent-4-enoic acid |
Specifications
| 16386-93-9 | |
| 108°C | |
| MFCD00012304 | |
| 3265 | |
| BGUAPYRHJPWVEM-UHFFFAOYSA-N | |
| 2,2-dimethylpent-4-enoic acid | |
| 140065 | |
| ≥98.0% (GC,T) | |
| 2,2-Dimethyl-4-pentenoic Acid |
| Yellow | |
| C7H12O2 | |
| 1 g | |
| 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g | |
| CC(C)(CC=C)C(O)=O | |
| 128.17 | |
| 128.17 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : No