missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-pentenoic Acid 98.0+%, TCI America™

Catalog No. D43541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43541G 1 g
D43545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43541G Supplier TCI America Supplier No. D43541G

Chemical Identifiers

CAS 16386-93-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00012304
InChI Key BGUAPYRHJPWVEM-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g
PubChem CID 140065
IUPAC Name 2,2-dimethylpent-4-enoic acid
SMILES CC(C)(CC=C)C(O)=O

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
UN Number 3265
Formula Weight 128.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-4-pentenoic Acid
EINECSNumber (2)-0609
TSCA No