missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-4-pentenoic acid, 95%

Catalog No. AAH6094006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6094006 5 g
AAH6094003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6094006 Supplier Thermo Scientific Chemicals Supplier No. H6094006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16386-93-9 | C7H12O2 | 128.17 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

Chemical Identifiers

CAS 16386-93-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00012304
InChI Key BGUAPYRHJPWVEM-UHFFFAOYSA-N
Synonym 2,2-dimethyl-4-pentenoic acid, 4-pentenoic acid, 2,2-dimethyl, 2,2-dimethyl-pent-4-enoic acid, 2,2-dimethyl-4-pentenoicacid, pubchem12797, acmc-1c5lb, ksc495e3r, 2,2-dimethyl4-pentenoic acid, 2,2-dimethyl-1-4-pentenoic acid 1g
PubChem CID 140065
IUPAC Name 2,2-dimethylpent-4-enoic acid
SMILES CC(C)(CC=C)C(O)=O

Specifications

Density 0.933
Boiling Point 104°C to 108°C
Flash Point 88°C (190°F)
Refractive Index 1.431
Linear Formula H2C=CHCH2C(CH3)2CO2H
Quantity 5 g
UN Number UN3265
Solubility Information Soluble in water.
Formula Weight 128.17
Percent Purity 95%
Chemical Name or Material 2,2-Dimethyl-4-pentenoic acid
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only