missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-3-butenoic acid, 95%

Catalog No. p-7055477
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6406703 1 g
AAH6406706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6406703 Supplier Thermo Scientific Chemicals Supplier No. H6406703
Only null left

CAS: 10276-09-2 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10276-09-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00196967
InChI Key SCFWAOWWAANBPY-UHFFFAOYSA-N
PubChem CID 139122
IUPAC Name 2,2-dimethylbut-3-enoic acid
SMILES CC(C)(C=C)C(=O)O

Specifications

Melting Point -6°C
Color Colorless
Density 0.957 g/mL
Boiling Point 185°C
Refractive Index 1.4295
Quantity 1 g
UN Number UN3265
Beilstein 1700758
Synonym 3-butenoic acid, 2,2-dimethyl, 2,2-dimethyl-but-3-enoic acid, 2,2-dimethyl-3-butenoic acid, ch2=chc ch3 2cooh, 3-butenoic acid,2,2-dimethyl
Formula Weight 114.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-3-butenoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only