missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine, 97%

Catalog No. p-7041108
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61058MD 250 mg
AAH6105803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61058MD Supplier Thermo Scientific Chemicals Supplier No. H61058MD
Only null left

CAS: 866089-28-3 | C10H13NO | 163.22 g/mol

It possess various biological activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It possess various biological activities.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

Chemical Identifiers

CAS 866089-28-3
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD11603419
InChI Key WILBVUMFTFLAHJ-UHFFFAOYSA-N
Synonym 2,2-dimethyl-3,4-dihydro-2h-1,4-benzoxazine, 2,2-dimethyl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine,3,4-dihydro-2,2-dimethyl, 2,2-dimethyl-3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-2,2-dimethyl
PubChem CID 23587101
IUPAC Name 2,2-dimethyl-3,4-dihydro-1,4-benzoxazine
SMILES CC1(CNC2=CC=CC=C2O1)C

Specifications

Quantity 250 mg
Formula Weight 163.22
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-3,4-dihydro-2H-1,4-benzoxazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only