missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC27201DA

Quantity 1g

 View more versions of this product

Catalog No. CC27201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

42327-95-7
192.214
AVVACPBRCFXZMR-UHFFFAOYSA-N
2795469
CC1(CC2=CC=CC(=C2O1)C(=O)O)C
C11H12O3
MFCD03426826
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carboxylic acid, 2, 3-dihydro-2, 2-dimethylbenzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carboxylic acid
2,2-dimethyl-3H-1-benzofuran-7-carboxylic acid

Specifications

2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-carboxylic acid
96.5
97%
C11H12O3
MFCD03426826
AVVACPBRCFXZMR-UHFFFAOYSA-N
2,2-dimethyl-3H-1-benzofuran-7-carboxylic acid
2795469
97%
42327-95-7
100.0
Amber glass bottle
1g
2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydrobenzofuran-7-carboxylic acid, 2, 3-dihydro-2, 2-dimethylbenzofuran-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-benzofuran-7-carboxylic acid, 7-benzofurancarboxylicacid, 2,3-dihydro-2,2-dimethyl, 2,2-dimethyl-2,3-dihydrobenzo b furan-7-carboxylic acid, 2,2-dimethyl-2,3-dihydro-1-benzo b furan-7-carboxylic acid
CC1(CC2=CC=CC(=C2O1)C(=O)O)C
192.214
192.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents