missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-2,3-dihydro-1-benzofuran-7-amine, 95%, Thermo Scientific™

Catalog No. p-3522431
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC27214CB 250mg Amber Glass Bottle
CC27214DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC27214CB Supplier Thermo Scientific Chemicals Supplier No. CC27214CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68298-46-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00075581
InChI Key LMTIGABGABPAGU-UHFFFAOYSA-N
Synonym 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-amine, unii-c3e03fdp2f, 2,3-dihydro-2,2-dimethyl-7-aminobenzofuran, 7-amino-2,3-dihydro-2,2-dimethylbenzofuran, 7-benzofuranamine, 2,3-dihydro-2,2-dimethyl, c3e03fdp2f, 2,3-dihydro-2,2-dimethylbenzofuran-7-amine, 2,2-dimethyl-2,3-dihydrobenzofuran-7-amine, 2,3-dihydro-2,2-dimethyl-7-benzofuranamine, dsstox_cid_24715
PubChem CID 91697
IUPAC Name 2,2-dimethyl-3H-1-benzofuran-7-amine
SMILES CC1(C)CC2=CC=CC(N)=C2O1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 163.22
Percent Purity 95%
Chemical Name or Material 2, 2-Dimethyl-2, 3-dihydro-1-benzofuran-7-amine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.