missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 408550025

 View more versions of this product

Catalog No. AC408550025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 2-Dimethyl-1, 3-propanediol
126-30-7
98.5
208.0°C
Authentic
Plastic bottle
HOCH2C(CH3)2CH2OH
2.5kg
05,259
neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
SLCVBVWXLSEKPL-UHFFFAOYSA-N
2,2-dimethylpropane-1,3-diol
31344
99%
99%
0.1mg KOH/g max.
100.0
107°C
98.5% min. (GC)
C5H12O2
MFCD00004685
01, IV, 2551
15, 6543
Solubility in water: 830g/L (20°C). Other solubilities: freely soluble in alcohol and ether
CC(C)(CO)CO
104.15
104.15

Chemical Identifiers

126-30-7
104.15
SLCVBVWXLSEKPL-UHFFFAOYSA-N
31344
CC(C)(CO)CO
C5H12O2
MFCD00004685
neopentyl glycol, 2,2-dimethyl-1,3-propanediol, dimethylolpropane, neopentylene glycol, neopentanediol, neopentylglycol, 1,3-propanediol, 2,2-dimethyl, neol, hydroxypivalyl alcohol, dimethyltrimethylene glycol
2,2-dimethylpropane-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 204-781-0

RTECSNumber : TY5775000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only