missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediamine 98.0+%, TCI America™

Catalog No. p-7126796
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
400 g
Catalog No. Quantity
D157025G 25 g
D1570400G 400 g
2 options

Catalog No. D157025G

Supplier: TCI America D157025G

Only null left

Chemical Identifiers

CAS 7328-91-8
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.181
MDL Number MFCD00009801
InChI Key DDHUNHGZUHZNKB-UHFFFAOYSA-N
PubChem CID 81770
IUPAC Name 2,2-dimethylpropane-1,3-diamine
SMILES CC(C)(CN)CN

Specifications

Color White
Boiling Point 65°C
Quantity 25 g
UN Number 2921
Synonym 2,2-dimethyl-1,3-propanediamine, neopentyldiamine, 1,3-propanediamine, 2,2-dimethyl, neopentanediamine, 1,3-diamino-2,2-dimethylpropane, 2,2-dimethyltrimethylenediamine, 1,3-propanediamine,2,2-dimethyl, neopentylenediamine, 2,3-propanediamine, acmc-209oqg
Formula Weight 102.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,2-Dimethyl-1,3-propanediamine