missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-propanediamine, 99%

Catalog No. p-4229188
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC210510500 50 g Glass bottle
AC210512500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210510500 Supplier Thermo Scientific Chemicals Supplier No. 210510500
Only null left

CAS: 7328-91-8 | C5H14N2 | 102.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7328-91-8
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
InChI Key DDHUNHGZUHZNKB-UHFFFAOYSA-N
PubChem CID 81770
IUPAC Name 2,2-dimethylpropane-1,3-diamine
SMILES CC(C)(CN)CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 29.0°C to 31.0°C
Color Colorless
Density 0.8500g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4566
Linear Formula H2NCH2C(CH3)2CH2NH2
Quantity 50 g
Beilstein 04,III,595
Specific Gravity 0.85
Synonym 2,2-dimethyl-1,3-propanediamine, neopentyldiamine, 1,3-propanediamine, 2,2-dimethyl, neopentanediamine, 1,3-diamino-2,2-dimethylpropane, 2,2-dimethyltrimethylenediamine, 1,3-propanediamine,2,2-dimethyl, neopentylenediamine, 2,3-propanediamine, acmc-209oqg
Solubility Information Solubility in water: miscible
Formula Weight 102.18
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2, 2-Dimethyl-1, 3-propanediamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
EINECSNumber 230-819-

RUO – Research Use Only