missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dimethyl-1,3-dioxolane-4-methanol 98.0+%, TCI America™
Supplier: TCI America D0710100G
Specifications
| 2,2-Dimethyl-1,3-dioxolane-4-methanol | |
| Colorless | |
| C6H12O3 | |
| 100 g | |
| RNVYQYLELCKWAN-UHFFFAOYSA-N | |
| (2,2-dimethyl-1,3-dioxolan-4-yl)methanol | |
| 7528 | |
| ≥98.0% (GC) |
| 100-79-8 | |
| 80°C | |
| MFCD00063238 | |
| solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal | |
| CC1(OCC(O1)CO)C | |
| 132.159 | |
| 132.16 | |
| Liquid |
Chemical Identifiers
| 100-79-8 | |
| 132.159 | |
| RNVYQYLELCKWAN-UHFFFAOYSA-N | |
| 7528 | |
| CC1(OCC(O1)CO)C |
| C6H12O3 | |
| MFCD00063238 | |
| solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal | |
| (2,2-dimethyl-1,3-dioxolan-4-yl)methanol |
Safety and Handling
EINECSNumber : (2)-2476&(5)-3554
RTECSNumber : JI0400000
TSCA : Yes