missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxolane-4-methanamine, 97%

Catalog No. AC397410050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397410050 5 g Glass bottle
AC397410010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397410050 Supplier Thermo Scientific Chemicals Supplier No. 397410050

CAS: 22195-47-7 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22195-47-7
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD01321384
InChI Key HXOYWCSTHVTLOW-UHFFFAOYNA-N
Synonym 2,2-dimethyl-1,3-dioxolane-4-methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-methylamine, 1-2,2-dimethyl-1,3-dioxolan-4-yl methanamine, 2,2-dimethyl-1,3-dioxolan-4-yl methylamine, 2,2-dimethyl-1,3-dioxolan-4-methanamine, 1,3-dioxolane-4-methanamine, 2,2-dimethyl, 2,2-dimethyl-4-aminomethyl-1,3-dioxolane, 2.2-dimethyl-1,3-dioxolane-4-methanamine
PubChem CID 4122349
IUPAC Name (2,2-dimethyl-1,3-dioxolan-4-yl)methanamine
SMILES CC1(C)OCC(CN)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0120g/mL
Boiling Point 164°C to 165°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.012
Formula Weight 131.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2-Dimethyl-1, 3-dioxolane-4-methanamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only