missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dioxan-5-one 97.0+%, TCI America™

Catalog No. D23971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23971G 1 g
D23975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23971G Supplier TCI America Supplier No. D23971G

Chemical Identifiers

CAS 74181-34-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00671514
InChI Key ASFQDNDZFGFMMP-UHFFFAOYSA-N
Synonym 2,2-dimethyl-1,3-dioxane-5-one, 2,2-dimethyl-1,3 dioxan-5-one, 1,3-dioxan-5-one, 2,2-dimethyl, acmc-1bg7h, ksc496q3n, 5-oxo-2,2-dimethyl-1,3-dioxane, 2,2-dimethyl-1,3-dioxan-5-one, technical 250mg
PubChem CID 2733141
IUPAC Name 2,2-dimethyl-1,3-dioxan-5-one
SMILES CC1(OCC(=O)CO1)C

Specifications

Color Yellow
Boiling Point 67°C
Quantity 1 g
UN Number 1993
Formula Weight 130.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-1,3-dioxan-5-one
TSCA No
Recommended Storage Refrigerator