missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-1,3-dichloropropane 95.0+%, TCI America™

Catalog No. D087025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D087025G 25 g
D08705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D087025G Supplier TCI America Supplier No. D087025G

Chemical Identifiers

CAS 29559-55-5
Molecular Formula C5H10Cl2
Molecular Weight (g/mol) 141.035
MDL Number MFCD00039363
InChI Key KTWNITKLQPCZSL-UHFFFAOYSA-N
Synonym 1,3-Dichloro-2,2-dimethylpropane, 1,3-Dichloroneopentane
PubChem CID 122408
IUPAC Name 1,3-dichloro-2,2-dimethylpropane
SMILES CC(C)(CCl)CCl

Specifications

Color Colorless
Boiling Point 138°C
Quantity 25 g
UN Number 1993
Formula Weight 141.04
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Dimethyl-1,3-dichloropropane