missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethoxy-2-phenylacetophenone 98.0+%, TCI America™

Catalog No. D170225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D170225G 25 g
D1702100G 100 g
D1702500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D170225G Supplier TCI America Supplier No. D170225G

Chemical Identifiers

CAS 24650-42-8
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.30
MDL Number MFCD00008475
InChI Key KWVGIHKZDCUPEU-UHFFFAOYSA-N
Synonym 2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
PubChem CID 90571
IUPAC Name 2,2-dimethoxy-1,2-diphenylethan-1-one
SMILES COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 169°C
Quantity 25 g
UN Number 3077
Formula Weight 256.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Dimethoxy-2-phenylacetophenone
EINECSNumber (4)-1357
TSCA Yes