missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoroethylamine 98.0+%, TCI America™

Catalog No. D47581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47581G 1 g
D47585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47581G Supplier TCI America Supplier No. D47581G

Chemical Identifiers

CAS 430-67-1
Molecular Formula C2H5F2N
Molecular Weight (g/mol) 81.066
MDL Number MFCD00798137
InChI Key OVRWUZYZECPJOB-UHFFFAOYSA-N
PubChem CID 136274
IUPAC Name 2,2-difluoroethanamine
SMILES C(C(F)F)N

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
UN Number 2733
Formula Weight 81.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoroethylamine