missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoroethyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7128727
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52991G 1 g
D52995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52991G Supplier TCI America Supplier No. D52991G
Only null left

Chemical Identifiers

CAS 74427-22-8
Molecular Formula C3H3F5O3S
Molecular Weight (g/mol) 214.106
MDL Number MFCD02093342
InChI Key NKULBUOBGILEAR-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2,2-Difluoroethyl Ester, 2,2-Difluoroethyl Triflate
PubChem CID 2774089
IUPAC Name 2,2-difluoroethyl trifluoromethanesulfonate
SMILES C(C(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 118°C
Quantity 1 g
UN Number 2920
Formula Weight 214.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Difluoroethyl Trifluoromethanesulfonate