missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluorocyclopropanemethanol, 97%

Catalog No. p-7031120
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32083MD 250 mg
AAH3208303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32083MD Supplier Thermo Scientific Chemicals Supplier No. H32083MD

CAS: 509072-57-5 | C4H6F2O | 108.088 g/mol

2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluorocyclopropanemethanol is used as an intermediate in the preparation of (2,2-difluorocyclopropyl)methylamine. It is also used as a reagent in the synthesis of pyrrolopyridine derivatives, which finds application as an antiviral agent.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 509072-57-5
Molecular Formula C4H6F2O
Molecular Weight (g/mol) 108.088
MDL Number MFCD06797748
InChI Key XOLSMTBBIZDHSG-UHFFFAOYSA-N
Synonym 2,2-difluorocyclopropyl methanol, 2,2-difluorocyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 97+%, soqdfiarbswxuup@, 2,2-difluoro-cyclopropanemethanol, 2,2-difluorocyclopropyl-methanol, 2,2-difluoro-cyclopropyl-methanol, cyclopropanemethanol, 2,2-difluoro
PubChem CID 2761401
IUPAC Name (2,2-difluorocyclopropyl)methanol
SMILES C1C(C1(F)F)CO

Specifications

Boiling Point 129°C
Quantity 250 mg
UN Number UN1987
Formula Weight 108.09
Percent Purity 97%
Chemical Name or Material 2,2-Difluorocyclopropanemethanol
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only