missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-N-methoxy-N-methylacetamide, 95%

Catalog No. AAH2583303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2583303 1 g
AAH2583306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2583303 Supplier Thermo Scientific Chemicals Supplier No. H2583303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 142492-01-1 | C4H7F2NO2 | 139.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 142492-01-1
Molecular Formula C4H7F2NO2
Molecular Weight (g/mol) 139.10
MDL Number MFCD06248152
InChI Key CUPRFYMJGQMIIC-UHFFFAOYSA-N
PubChem CID 19023731
IUPAC Name 2,2-difluoro-N-methoxy-N-methylacetamide
SMILES CON(C)C(=O)C(F)F

Specifications

Boiling Point 40°C to 41°C (0.7 mmHg)
Refractive Index 1.402
Quantity 1 g
Synonym n-methyl-n-methoxydifluoroacetamide, acetamide, 2,2-difluoro-n-methoxy-n-methyl, acmc-20a3nv, n-methoxy-n-methyldifluoroacetamide, n-methoxy-n-methyl difluoroacetamide, n-difluoroacetyl-n-methoxymethylamine, acetamide,2,2-difluoro-n-methoxy-n-methyl, 2,2-bis fluoranyl-n-methoxy-n-methyl-ethanamide
Formula Weight 139.1
Percent Purity 95%
Chemical Name or Material 2,2-Difluoro-N-methoxy-N-methylacetamide
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only