missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 656-46-2
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.12
InChI Key VJLDRFCNFNQTTH-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
PubChem CID 608772
IUPAC Name 2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
SMILES C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
View More Specs

Products 1
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
missing translation for 'productCode' missing translation for 'brand' Quantity Packaging Price Quantity & Availability  
AC449140050
View Documents Promotion Details
N/A 5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

656-46-2
202.12
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
C8H4F2O4
VJLDRFCNFNQTTH-UHFFFAOYSA-N
608772
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F

Specifications

656-46-2
C8H4F2O4
VJLDRFCNFNQTTH-UHFFFAOYSA-N
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid
608772
97%
96% min. (HPLC)
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F
202.12
202.12
2, 2-Difluoro-1, 3-benzodioxole-5-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only