missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde, 97%

Catalog No. AAB2423203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2423203 1 g
AAB2423206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2423203 Supplier Thermo Scientific Chemicals Supplier No. B2423203

CAS: 656-42-8 | C8H4F2O3 | 186.114 g/mol

2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde is used as primary and secondary intermediate.

Solubility
Sparingly soluble in water (0.31 g/L at 25°C).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air

Chemical Identifiers

CAS 656-42-8
Molecular Formula C8H4F2O3
Molecular Weight (g/mol) 186.114
MDL Number MFCD00792420
InChI Key GGERGLKEDUUSAP-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole-5-carbaldehyde, 2,2-difluorobenzodioxole-5-carboxaldehyde, 2,2-difluoro-1,3-benzodioxole-5-carboxaldehyde, 2,2-difluoro-2h-1,3-benzodioxole-5-carbaldehyde, 2,2-difluoro-5-formylbenzodioxole, 2,2-difluoro-5-formyl-1,3-benzodioxole, 2,2-difluoro-benzo 1,3 dioxole-5-carbaldehyde, 5-formyl-2,2-difluorobenzodioxole, 1,3-benzodioxole-5-carboxaldehyde, 2,2-difluoro, 2,2-difluoro-1,3-benzodioxole-5-arboxaldehyde
PubChem CID 2736973
IUPAC Name 2,2-difluoro-1,3-benzodioxole-5-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)OC(O2)(F)F

Specifications

Density 1.399
Boiling Point 103°C to 105°C (20 mmHg)
Refractive Index 1.499
Quantity 1 g
Beilstein 1369253
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water (0.31g/L at 25°C).
Formula Weight 186.11
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-5-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only