missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid, 97%

Catalog No. p-7027302
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2395503 1 g
AAB2395506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2395503 Supplier Thermo Scientific Chemicals Supplier No. B2395503

CAS: 126120-85-2 | C8H4F2O4 | 202.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126120-85-2
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.113
MDL Number MFCD01631473
InChI Key ZGAQVJDFFVTWJK-UHFFFAOYSA-N
Synonym 2,2-difluorobenzo d 1,3 dioxole-4-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-4-carboxylic acid, 2,2-difluoro-1,3-benzodioxole-4-carboxylicacid, 2,2-difluoro-benzo 1,3 dioxole-4-carboxylic acid, 2,2-difluorobenzo d 1,3-dioxolene-4-carboxylic acid, acmc-1c3im, 1,3-benzodioxole-4-carboxylicacid, 2,2-difluoro, 2,2-bis fluoranyl-1,3-benzodioxole-4-carboxylic acid, 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
PubChem CID 2774067
IUPAC Name 2,2-difluoro-1,3-benzodioxole-4-carboxylic acid
SMILES C1=CC(=C2C(=C1)OC(O2)(F)F)C(=O)O

Specifications

Melting Point 202°C to 205°C
Quantity 1 g
Beilstein 7292629
Formula Weight 202.12
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-1,3-benzodioxole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only