missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(Diethylamino)ethoxy]ethanol 98.0+%, TCI Americaâ„¢
$58.59 - $544.25
Chemical Identifiers
CAS | 140-82-9 |
---|---|
Molecular Formula | C8H19NO2 |
Molecular Weight (g/mol) | 161.25 |
MDL Number | MFCD00020604 |
InChI Key | VKBVRNHODPFVHK-UHFFFAOYSA-N |
Synonym | Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether |
PubChem CID | 67327 |
IUPAC Name | 2-[2-(diethylamino)ethoxy]ethan-1-ol |
SMILES | CCN(CC)CCOCCO |
Chemical Identifiers
140-82-9 | |
161.25 | |
VKBVRNHODPFVHK-UHFFFAOYSA-N | |
67327 | |
CCN(CC)CCOCCO |
C8H19NO2 | |
MFCD00020604 | |
Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether | |
2-[2-(diethylamino)ethoxy]ethan-1-ol |
Specifications
140-82-9 | |
101°C | |
MFCD00020604 | |
Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether | |
CCN(CC)CCOCCO | |
161.25 | |
161.25 | |
Liquid |
Yellow | |
C8H19NO2 | |
25 g | |
VKBVRNHODPFVHK-UHFFFAOYSA-N | |
2-[2-(diethylamino)ethoxy]ethan-1-ol | |
67327 | |
≥98.0% (GC) | |
2-[2-(Diethylamino)ethoxy]ethanol |
Safety and Handling
EINECSNumber : (9)-0562
TSCA : Yes