missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Diethylamino)ethoxy]ethanol 98.0+%, TCI America™

Catalog No. D029825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D029825G 25 g
D0298500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D029825G Supplier TCI America Supplier No. D029825G
Only null left

Chemical Identifiers

CAS 140-82-9
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.25
MDL Number MFCD00020604
InChI Key VKBVRNHODPFVHK-UHFFFAOYSA-N
Synonym Ethylene Glycol Mono[2-(diethylamino)ethyl] Ether
PubChem CID 67327
IUPAC Name 2-[2-(diethylamino)ethoxy]ethan-1-ol
SMILES CCN(CC)CCOCCO

Specifications

Color Yellow
Boiling Point 101°C
Quantity 25 g
Formula Weight 161.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-[2-(Diethylamino)ethoxy]ethanol
EINECSNumber (9)-0562
TSCA Yes